WHA10463814
Whatman® Roby automated filter 25 mm
glass fiber pre-filter GF55, pore size 0.7 μm
Synonym(s):
Z751723
About This Item
Recommended Products
material
circles
glass fiber membrane
polypropylene housing
sterility
non-sterile
packaging
pk of 200
manufacturer/tradename
Whatman 10463814
Whatman Article No. 28415886 (US reference)
diam.
25 mm
pore size
0.7 μm
Looking for similar products? Visit Product Comparison Guide
General description
The filter housing is made from mechanically stable polypropylene. The external geometry of the filter housing ensures simple and smooth filter transport from the storage turntable to the filtration site and easy filter changing.
Features
Optimized for Sotax, Caliper, Zymark® and Varian® tablet testers
Mechanically stable polypropylene
Benefits
Easy filter changing
Ensures simple and smooth filter transport
Applications
Fine filtration of samples in the automatic tablet dissolution test
Method development with the Roby 25 Filter Validation Kit
Suitability
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service