WHA10463423
Whatman® UNIFLO® 25 syringe filters
PTFE membrane, pore size 0.45 μm, pkg of 1000
Synonym(s):
Whatman syringe filter, Z761370, filtration, syringe filters
About This Item
Recommended Products
material
PTFE membrane
polypropylene housing (housing)
polypropylene housing
sterility
non-sterile
packaging
pkg of 1000
manufacturer/tradename
Whatman 10463423
Whatman Article No. 28415822 (US reference)
diam.
25 mm
pore size
0.45 μm
fitting
female inlet fitting for Luer Lock
male outlet fitting for Slip Luer
suitability
suitable for (volumes of 100mL or less; filter provides easy separation of solutions)
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- 13 or 25 mm diameters
- 0.2 μm, 0.22 μm or 0.45 μm pore sizes
- Sterile or non-sterile options
- Individual printing on the filter for easy identification
- Bench top space saving packaging
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service