Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WHA10462257

Whatman® Celtron 30/0.45

45 CA-GF92

Synonym(s):

Z751499

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104922
NACRES:
NB.24

material

cellulose acetate membrane
polypropylene housing

sterility

sterile

feature

glass fiber prefilter

packaging

pk of 50

manufacturer/tradename

Whatman 10462257
Whatman Article No. 28415743 (US reference)

diam.

30 mm

volume

≤100 mL

pore size

0.45 μm

Looking for similar products? Visit Product Comparison Guide

General description

For difficult-to-filter solutions Celtron ready-to-use filter units have an integral glass fiber prefilter to retain coarse particles and a membrane filter for filtering the fine contaminants. This filter combination increases the throughput with less filtration force being required.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service