Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WHA10416304

Whatman® Nytran SuPerCharge (SPC) TurboBlotter

W × L 11 cm × 14 cm, pore size 0.45 μm, pkg of 1 ea

Synonym(s):

Z698091, blotting membrane, nylon membranes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

packaging

pkg of 1 ea

manufacturer/tradename

Whatman 10416304
Whatman Article No. 28415383 (US reference)

W × L

11 cm × 14 cm

pore size

0.45 μm

suitability

suitable for (high-resolution transfer of DNA and RNA; nucleic acid transfer)

Looking for similar products? Visit Product Comparison Guide

General description


The TurboBlotter system is a rapid downward blotting device for high-resolution transfer of DNA and RNA.

The conventional Northern/Southern transfer stack has been turned upside down in order to take advantage of gravity. No heavy weights are required on the top of the transfer stack, eliminating the messy set-up of standard upward capillary transfers.

The TurboBlotter System offers greater speed, target resolution, and convenience versus traditional blotting procedures. Alkaline DNA transfers can be performed in as little as one hour, while neutral (SSC) transfers of DNA and RNA take only three hours.

TurboBlotter is available in two sizes. The smaller model is 12 x 16 cm, for gels from 7 x 8 cm to 11 x 14 cm. The larger model is 21 x 26 cm, for gels from 12 x 21 cm to 20 x 25 cm. Each size comes in your choice of refill packs and the refill packs are available separately.

Features and Benefits:

  • Rapid - downward capillary transfer allows for alkaline buffer transfers in one hour and neutral (SSC) transfers in three hours
  • Economical - reusable blotting device requires less buffer and blotting paper. Convenient refill packs available
  • Compact - has a smaller footprint than most homemade devices and is stackable. Up to five units can be stacked on top of each other during transfers
  • Easy to use - very easy to set up and works without power or vacuum source
  • Uses Whatman Nytran SPC membrane and Whatman 3MM Chr and GB004 Blotting Papers for top performance

Each transfer uses one sheet of Nytran SPC, one wick of 3MM Chr, and a blotter stack composed of eight sheets of 3MM Chr and twenty sheets of GB004. Each TurboBlotter Kit or TurboBlotter refill contains five sets of these materials, enough for five transfers.

TurboBlotter Kits, Small
12 x 16 cm transfer device with Nytran SPC, wicks and blotter stacks for indicated gel size.

TurboBlotter Refills, Small
Nytran SPC, wicks and blotter stacks for five transfers from indicated gel size.

TurboBlotter Kits, Large
21 x 26 cm transfer device with Nytran SPC, wicks and blotter stacks for indicated gel size.

TurboBlotter Refills, Large
Nytran SPC, wicks and blotter stacks for five transfers from indicated gel size.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Nytran is a trademark of Cytiva
Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service