Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W778567

Sigma-Aldrich

L-Cysteine hydrochloride

FG, product of Wacker Chemie AG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NH2)COOH · HCl
CAS Number:
Molecular Weight:
157.62
Beilstein/REAXYS Number:
3560277
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Flavis number:
17.032
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal

reg. compliance

EU Regulation 178/2002

optical activity

[α]/D 5.7 to 6.8° (Ph. Eur. (2.2.7))

impurities

≤10 ppm heavy metals (Ph. Eur. (2.4.8/A))
≤200 ppm ammonium (Ph. Eur. (2.4.1B))

ign. residue

≤0.1% (Ph. Eur. (2.4.14))

loss

≤2.0% loss on drying

solubility

H2O: 250 mg/10 mL, clear, colorless

cation traces

As: ≤1 ppm (Ph. Eur. (2.4.2/A))
Pb: ≤5 ppm

suitability

clear, colorless for appearance of solution (Ph. Eur. (2.2.1))
positive for identity (Ph. Eur. (2.2.24))

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

odorless

SMILES string

Cl[H].N[C@@H](CS)C(O)=O

InChI

1S/C3H7NO2S.ClH/c4-2(1-7)3(5)6;/h2,7H,1,4H2,(H,5,6);1H/t2-;/m0./s1

Inchi Key

IFQSXNOEEPCSLW-DKWTVANSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-cysteine is a sulfur-containing amino acid.[1]

Application

The addition of L-cysteine hydrochloride to wheat flour led to the reduction in the peak viscosity during the analysis of its pasting characteristics using rapid visco analyzer (RVA).[2]

Biochem/physiol Actions

NMDA glutamatergic receptor agonist that is also an agonist at AMPA glutamatergic receptors at high concentrations.
NMDA glutamatergic receptor agonist; AMPA glutamatergic receptor agonist at high concentrations.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of Rapid Visco Analyser (RVA) for measuring the pasting characteristics of wheat flour as influenced by additives.
Ravi R, et al
Journal of the Science of Food and Agriculture, 79(12), 1571-1576 (1999)
Antioxidant effects of sulfur-containing amino acids
Atmaca G
Yonsei Medical Journal, 45, 776-788 (2004)
Chen Ding et al.
Microbial ecology, 70(2), 433-444 (2015-03-19)
Anaerobic alkane degradation via methanogenesis has been intensively studied under mesophilic and thermophilic conditions. While there is a paucity of information on the ability and composition of anaerobic alkane-degrading microbial communities under low temperature conditions. In this study, we investigated
Sabine A Tanner et al.
The British journal of nutrition, 114(5), 746-755 (2015-08-28)
Modulating the gut microbiota via dietary interventions is a common strategy to enhance the natural defence mechanisms of the host. Several in vitro studies have highlighted the probiotic potential of Bifidobacterium thermophilum RBL67 (RBL67) selected for its anti-Salmonella effects. The
Min Hyung Kang et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 87(3), 489-499 (2014-04-08)
To facilitate selective drug delivery to hepsin (Hpn)-expressing cancer cells, the RIPL peptide (IPLVVPLRRRRRRRRC; 16mer; 2.1 kDa) was synthesized as a novel cell penetrating/homing peptide (CPHP) and conjugated to a liposomal carrier. RIPL peptide-conjugated liposomes (RIPL-Lipo) were prepared by conjugating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service