Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W550701

Sigma-Aldrich

2-Methyl-3-tetrahydrofuranthiol solution

1.0 wt. % in propylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.16
organoleptic:
sulfurous
biological source:
synthetic
Pricing and availability is not currently available.

biological source

synthetic

concentration

1.0 wt. % in propylene glycol

refractive index

n20/D 1.4330

bp

187 °C

density

1.036 g/mL at 25 °C

application(s)

flavors and fragrances

organoleptic

sulfurous

SMILES string

CC1OCCC1S

InChI

1S/C5H10OS/c1-4-5(7)2-3-6-4/h4-5,7H,2-3H2,1H3

InChI key

DBPHPBLAKVZXOY-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

225.0 °F - closed cup

flash_point_c

107.2 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service