Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W530618

Sigma-Aldrich

Coffee oil

roasted, natural (US), FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12164502
NACRES:
NA.21

grade

FG
Halal
natural (US)

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 173.255
FDA 21 CFR 173.290
FDA 21 CFR 182.20

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

coffee; roasted

Looking for similar products? Visit Product Comparison Guide

Application

This coffee preparation will add roasted character to all coffee flavors and blends. It will add a dark note to chocolate and cocoa flavors. It can also be used in flavors requiring roasted or burnt notes like bacon, nut and toasted marshmallow.[1]

Biochem/physiol Actions

Taste at 5-10ppm[1]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

>199.9 °F

flash_point_c

> 93.3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Michalski, J.
Perfumer & Flavorist, 39(4), 52-52 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service