Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W530427

Sigma-Aldrich

Nutmeg extract

natural

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12164502

biological source

Myristica fragrans Houtt.

grade

Kosher
natural

composition

Limonene, 4.7%
Linalool, 0.4%
Methyl eugenol, 0.9%

refractive index

n20/D 1.499

density

0.963 g/mL at 25 °C

application(s)

flavors and fragrances

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable Liquids

wgk_germany

WGK 2

flash_point_f

109.4 °F - closed cup

flash_point_c

43 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service