Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W530165

Sigma-Aldrich

Evening primrose oil

Certified organic (NOP)

Synonym(s):

Oenothera biennis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164502

biological source

Oenothera biennis

grade

Certified organic (NOP)
Halal
Kosher

origin

China origin

refractive index

n20/D 1.479

density

0.926 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

floral

Looking for similar products? Visit Product Comparison Guide

Legal Information

Certified organic by MOSA

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service