Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W530106

Sigma-Aldrich

Lime oil

Certified organic (NOP)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2631
MDL number:
UNSPSC Code:
12164502

grade

Certified organic (NOP)
Halal
Kosher

origin

Mexico origin

refractive index

n20/D 1.474-1.477 (lit.)
n20/D 1.476 (lit.)

density

0.861 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

lime

Looking for similar products? Visit Product Comparison Guide

General description

Cold pressed

Legal Information

Certified organic by MOSA

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

120.0 °F - closed cup

flash_point_c

48.9 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service