Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W526304

Sigma-Aldrich

Ethyl trans-4-octenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

grade

Kosher

contains

(may contain up to 0.10% alpha-tocopherol, synthetic as stabilzer)

refractive index

n20/D 1.432 (lit.)

bp

150 °C/11 mmHg (lit.)

density

0.878 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

ethereal

SMILES string

[H]\C(CCC)=C(\[H])CCC(=O)OCC

InChI

1S/C10H18O2/c1-3-5-6-7-8-9-10(11)12-4-2/h6-7H,3-5,8-9H2,1-2H3/b7-6+

InChI key

WRUZCQAJIHSQPL-VOTSOKGWSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

174.9 °F - closed cup

flash_point_c

79.4 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service