Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W524301

Sigma-Aldrich

Vetiver acetate, Java

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
organoleptic:
sweet
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

origin

USA origin

refractive index

n20/D 1.524 (lit.)

bp

286 °C (lit.)

density

0.976 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

sweet

SMILES string

CC1CC(OC(C)=O)C=C(C)C2C\C(CC12)=C(\C)C

InChI

1S/C17H26O2/c1-10(2)14-8-16-11(3)6-15(19-13(5)18)7-12(4)17(16)9-14/h6,12,15-17H,7-9H2,1-5H3

InChI key

UAVFEMBKDRODDE-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service