Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W523518

Sigma-Aldrich

Fir needle oil,Canadian

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164502

biological source

Abies balsamea

grade

Kosher

optical activity

[α]20/D −20.5°, neat

origin

Canada origin

refractive index

n20/D 1.4750 (lit.)

density

0.874 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

organoleptic

woody

Preparation Note

Extraction method: steam distillation

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

109.9 °F - closed cup

flash_point_c

43.3 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service