Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W521000

Sigma-Aldrich

Cade oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Juniperus oxycedrus

grade

Kosher

origin

Spain origin

refractive index

n20/D 1.5340 (lit.)

bp

184 °C (lit.)

density

1.027 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

organoleptic

smoky

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Extraction method: steam distillation and rectification

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

136.0 °F - closed cup

flash_point_c

57.8 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Zampetti et al.
The Journal of dermatological treatment, 19(3), 134-140 (2008-06-24)
Psoriasis requires lifelong treatments that depend on the extent, clinical forms and associated conditions. To retrospectively analyze which topical treatments were used, their efficacy, and potential advantages and disadvantages. A total of 666 patients admitted for the first time over
Prescrire international, 18(104), 275-275 (2009-12-24)
(1) Plaque psoriasis is the most common form of psoriasis in children. Topical agents should be tried first, especially well-tolerated products such as emollients. Topical corticosteroids are sometimes useful during exacerbations but, given adverse effects, they should only be used
B Schoket et al.
The Journal of investigative dermatology, 94(2), 241-246 (1990-02-01)
Preparations of coal-tar and juniper tar (cade oil) that are used in the treatment of psoriasis are known to contain numerous potentially carcinogenic polycyclic aromatic hydrocarbons (PAH). Evidence of covalent binding to DNA by components of these mixtures was sought
Grant T McQuate et al.
Journal of economic entomology, 97(3), 862-870 (2004-07-29)
Cade oil, a commercially available essential oil produced by destructive distillation of juniper, Juniperus oxycedrus L., twigs, is known to synergize the attractancy of alpha-ionol to male Bactrocera latifrons (Hendel). Through chemical fractionation and outdoor olfactometer-based bioassays, seven compounds in
G T McQuate et al.
Journal of economic entomology, 94(1), 39-46 (2001-03-10)
Male lures are known for many tephritid fruit fly species and are often preferred over food bait based traps for detection trapping because of their high specificity and ability to attract flies over a wide area. Alpha-ionol has been identified

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service