Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W516503

Sigma-Aldrich

Tetrahydromyrcenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)(CH2)3C(CH3)2OH
CAS Number:
Molecular Weight:
158.28
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

refractive index

n20/D 1.4335 (lit.)

bp

197-201 °C (lit.)

density

0.823 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

organoleptic

lime

SMILES string

CCC(C)CCCC(C)(C)O

InChI

1S/C10H22O/c1-5-9(2)7-6-8-10(3,4)11/h9,11H,5-8H2,1-4H3

InChI key

WRFXXJKURVTLSY-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

185.0 °F - closed cup

flash_point_c

85 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service