Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W510408

Sigma-Aldrich

Ethyl (±)-2-hydroxycaproate

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(OH)CO2C2H5
CAS Number:
Molecular Weight:
160.21
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
organoleptic:
green; fruity
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

assay

≥97%

refractive index

n20/D 1.425 (lit.)

bp

80-83 °C/11 mmHg (lit.)

density

0.967 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

green; fruity

SMILES string

CCCCC(O)C(=O)OCC

InChI

1S/C8H16O3/c1-3-5-6-7(9)8(10)11-4-2/h7,9H,3-6H2,1-2H3

InChI key

MRYSSTRVUMCKKB-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

208.4 °F - closed cup

flash_point_c

98 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service