Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W509205

Sigma-Aldrich

(S)-(−)-β-Citronellol

99%, FG

Synonym(s):

β-Rhodinol, (S)-3,7-Dimethyl-6-octen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
156.27
FEMA Number:
2309
Beilstein/REAXYS Number:
1721505
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.229
organoleptic:
geranium
grade:
FG
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012

vapor pressure

~0.02 mmHg ( 25 °C)

assay

99%

optical activity

[α]20/D −4.5°, neat

refractive index

n20/D 1.456 (lit.)

bp

225-226 °C (lit.)

density

0.856 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

geranium

SMILES string

C[C@H](CCO)CC\C=C(\C)C

InChI

1S/C10H20O/c1-9(2)5-4-6-10(3)7-8-11/h5,10-11H,4,6-8H2,1-3H3/t10-/m0/s1

InChI key

QMVPMAAFGQKVCJ-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

213.8 °F - DIN 51758

flash_point_c

101 °C - DIN 51758

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wantida Chaiyana et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(8-9), 836-839 (2012-04-19)
Inhibition of cholinesterase has attracted much attention recently because of its potential for the treatment of Alzheimer's disease. In this work, the anticholinesterase activities of plant oils were investigated using Ellman's colorimetric method. The results indicate that essential oils obtained
Renan G Brito et al.
Journal of natural medicines, 66(4), 637-644 (2012-02-22)
We describe the antinociceptive and anti-inflammatory properties of citronellol (CT) in rodents. CT, a monoterpene alcohol, is a naturally occurring monoterpene compound prevalent in essential oils of various aromatic plant species, such as Cymbopogon citratus. In mice, when evaluated against
Yoshikazu Kitano et al.
Biofouling, 27(2), 201-205 (2011-02-01)
Twenty novel simple alkyl isocyanides derived from citronellol were synthesized and evaluated for their antifouling activity and toxicity against cypris larvae of the barnacle, Balanus amphitrite. The anti-barnacle activity of the synthesized isocyanides was in the EC(50) range of 0.08-1.49
Hyland Cronin et al.
Journal of cosmetic dermatology, 9(3), 218-225 (2010-10-05)
New developments in the realm of skin rejuvenation such as phytotherapy are at an astounding increasing pace in the cosmeceutical market. Yet, many of these products that are classified as cosmeceuticals are tested less vigorously and do not have to
Shalinder Kaur et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 66(5-6), 260-266 (2011-08-05)
Citronellol, an oxygenated monoterpene, is found naturally in the essential oils of several aromatic plants and has been reported to exhibit growth inhibitory and pesticidal activities. However, its mechanism of action is largely unexplored. We investigated the effect of citronellol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service