Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W507725

Sigma-Aldrich

3-Ethoxypropionaldehyde diethyl acetal

≥95%

Synonym(s):

1,1,3-Triethoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH2CH(OC2H5)2
CAS Number:
Molecular Weight:
176.25
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.097
NACRES:
NA.21

biological source

synthetic

reg. compliance

FDA 21 CFR 117

assay

≥95%

refractive index

n20/D 1.406 (lit.)

bp

184-186 °C (lit.)

density

0.898 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

faint

SMILES string

CCOCCC(OCC)OCC

InChI

1S/C9H20O3/c1-4-10-8-7-9(11-5-2)12-6-3/h9H,4-8H2,1-3H3

InChI key

LGICWIVABSMSDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

127.4 °F - closed cup

flash_point_c

53 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service