Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W504718

Sigma-Aldrich

Azodicarboxamide

99%, FCC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CON=NCONH2
CAS Number:
Molecular Weight:
116.08
Beilstein/REAXYS Number:
1704003
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

grade

Kosher

reg. compliance

FCC
FDA 21 CFR 172.806
FDA 21 CFR 177.1210
FDA 21 CFR 177.2600
FDA 21 CFR 178.3010

assay

99%

mp

220-225 °C (dec.) (lit.)

application(s)

flavors and fragrances

organoleptic

odorless

SMILES string

NC(=O)\N=N\C(N)=O

InChI

1S/C2H4N4O2/c3-1(7)5-6-2(4)8/h(H2,3,7)(H2,4,8)/b6-5+

InChI key

XOZUGNYVDXMRKW-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

supp_hazards

Storage Class

4.1A - Other explosive hazardous materials

wgk_germany

WGK 1

flash_point_f

>392.0 °F

flash_point_c

> 200 °C

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaoru Midorikawa et al.
Free radical research, 36(6), 667-675 (2002-08-16)
Free radicals generated by chemicals can cause sequence-specific DNA damage and play important roles in mutagenesis and carcinogenesis. Carbamoyl group (CONH2) and its derived groups (CONR2) occur as natural products and synthetic chemical compounds. We have investigated the DNA damage
A S Pereira et al.
Food additives and contaminants, 21(1), 63-69 (2004-01-28)
Data from the Brazilian Agricultural Ministry show that before the implementation of the Brazilian programme of nitrofuran control in February 2003, the cases of contamination of Brazilian chicken by nitrofurans were almost exclusively due to furaltadone. After May 2003, such
M Hsu et al.
Journal of human virology, 3(1), 16-26 (2000-04-25)
An early step in the life cycle of human immunodeficiency virus type 1 is the reverse transcription of the viral RNA genome into double-stranded DNA, which is subsequently translocated to the cell nucleus. It is then integrated into host DNA
Hyeon Yoon et al.
Journal of biomaterials science. Polymer edition, 21(2), 159-170 (2010-01-23)
To study cell responses, polymeric scaffolds with a controllable pore size and porosity have been fabricated using rapid-prototyping methods. However, the scaffolds fabricated by rapid prototyping have very smooth surfaces, which tend to discourage initial cell attachment. Initial cell attachment
P P J Mulder et al.
Analytica chimica acta, 586(1-2), 366-373 (2007-03-28)
Recently doubts have arisen on the usefulness of semicarbazide as marker residue for the illegal use of the antibiotic nitrofurazone (NFZ) in aquaculture and poultry production. Most notably azodicarbonamide (ADC) has been implicated as an alternative source of semicarbazide. ADC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service