Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W503509

Sigma-Aldrich

2-Acetylthiophene

≥98%, FG

Synonym(s):

Methyl-2-thienyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6OS
CAS Number:
Molecular Weight:
126.18
Beilstein/REAXYS Number:
107910
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.040
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

assay

≥98%

refractive index

n20/D 1.565 (lit.)

bp

214 °C (lit.)

mp

10-11 °C (lit.)

density

1.168 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

organoleptic

sulfurous

SMILES string

CC(=O)c1cccs1

InChI

1S/C6H6OS/c1-5(7)6-3-2-4-8-6/h2-4H,1H3

InChI key

WYJOVVXUZNRJQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

206.6 °F

flash_point_c

97 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinbing Liu et al.
Bioorganic & medicinal chemistry, 16(3), 1096-1102 (2008-03-08)
A series of 1-(1-arylethylidene)thiosemicarbazide compounds and their analogues were synthesized and characterized by 1H NMR, MS. Their tyrosinase inhibitory activities were investigated by an assay based on the catalyzing ability of tyrosinase for the oxidation of L-DOPA, comparing with 4-methoxycinnamic
Mostafa M Ghorab et al.
Acta pharmaceutica (Zagreb, Croatia), 64(4), 419-431 (2014-12-23)
A novel series of thiophenes having biologically active sulfonamide 2-11, 3-methylisoxazole 12, 4-methoxybenzo[d] thiazole 13, quinoline 14, 15, benzoylphenylamino 16, and anthracene-9,10-dione 17 moieties were prepared. Structures of the newly synthesized compounds were established by elemental analysis and spectral data.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service