Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W442800

Sigma-Aldrich

Benzyl isothiocyanate

98%, FG

Synonym(s):

BITC, Benzyl mustard oil, Isothiocyanotaomethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NCS
CAS Number:
Molecular Weight:
149.21
FEMA Number:
4428
Beilstein/REAXYS Number:
386135
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.102
NACRES:
NA.21
organoleptic:
medicinal
grade:
FG
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012

assay

98%

refractive index

n20/D 1.601 (lit.)

bp

242-243 °C (lit.)

density

1.125 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

medicinal

storage temp.

2-8°C

SMILES string

S=C=NCc1ccccc1

InChI

1S/C8H7NS/c10-7-9-6-8-4-2-1-3-5-8/h1-5H,6H2

InChI key

MDKCFLQDBWCQCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl isothiocyanate occurs naturally in papaya seeds, moringa, and cruciferous vegetables.[1] It shows potent anti-proliferative activity against human pancreatic cancer cells.[2]

Application

Possible applications: Mustard, horseradish, vegetative nuances for cabbage, radish and watercress, tropical papaya mouthfeel effects.[3]

Biochem/physiol Actions

Odor at 1.0%: Vegetative cabbage and radishlike, green with a musty sulfurous earthiness reminiscent of mustard greens.
Taste at 0.5ppm: Green vegetative cabbage, kimchi, radish and wasabi-like with green leafy nuances and imparting a strong trigeminal tongue numbing and raspy sensation.[3]

Other Notes

Natural occurrence: watercress, nasturtium, papaya and endive[3]

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

222.8 °F

flash_point_c

106 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzyl isothiocyanate of papaya fruit.
Tang CS.
Phytochemistry, 10(1), 117-121 (1971)
Anuradha Sehrawat et al.
Cancer prevention research (Philadelphia, Pa.), 4(7), 1107-1117 (2011-04-06)
We showed previously that cruciferous vegetable constituent benzyl isothiocyanate (BITC) inhibits growth of cultured and xenografted human breast cancer cells and suppresses mammary cancer development in a transgenic mouse model. We now show, for the first time, that BITC inhibits
Wei-Ju Lee et al.
Journal of the science of food and agriculture, 91(13), 2348-2354 (2011-05-19)
In general, about 300 g kg(-1) of the weight of papaya fruits appears as waste materials during processing, including a considerable amount of papaya seeds. To make a more efficient use of papaya, it is worth investigating the utilization of
Younghyun Lee et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(7), 2397-2406 (2012-04-25)
Cis-diamminedichloroplatinum (II) (cisplatin) is one of the most widely used chemotherapeutic drugs, but its effectiveness is limited by tumor cell resistance and the severe side effects it causes. One strategy for overcoming this problem is the concomitant use of natural
Huiqin Yan et al.
The British journal of nutrition, 106(12), 1779-1791 (2011-07-08)
Dietary isothiocyanates have been shown to possess anti-tumour activity, inhibiting several types of cultured human cancer cell growth. However, there are limited studies on their effects on cancer cell metastasis. Our previous study showed that benzyl isothiocyanate (BITC) and phenethyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service