Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W435100

Sigma-Aldrich

5-Hexen-1-ol

98%

Synonym(s):

5-Hexenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2(CH2)3CH=CH2
CAS Number:
Molecular Weight:
100.16
FEMA Number:
4351
Beilstein/REAXYS Number:
1236458
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
organoleptic:
green
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

assay

98%

refractive index

n20/D 1.435 (lit.)

bp

78-80 °C/25 mmHg (lit.)

density

0.834 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

green

SMILES string

OCCCCC=C

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h2,7H,1,3-6H2

InChI key

UIZVMOZAXAMASY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

138.2 °F

flash_point_c

59 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service