Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W433300

Sigma-Aldrich

1-Pentanethiol

98%

Synonym(s):

Amyl mercaptan, Mercaptan C5, Pentyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4SH
CAS Number:
Molecular Weight:
104.21
FEMA Number:
4333
Beilstein/REAXYS Number:
1730979
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.191

biological source

synthetic

vapor pressure

27.4 mmHg ( 37.7 °C)

assay

98%

refractive index

n20/D 1.446 (lit.)

bp

126 °C (lit.)

density

0.84 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

alliaceous

SMILES string

CCCCCS

InChI

1S/C5H12S/c1-2-3-4-5-6/h6H,2-5H2,1H3

InChI key

ZRKMQKLGEQPLNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

64.4 °F - closed cup

flash_point_c

18 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service