Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W425900

Sigma-Aldrich

1-Heptanethiol

98%

Synonym(s):

Heptyl mercaptan, Mercaptan C7

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6SH
CAS Number:
Molecular Weight:
132.27
FEMA Number:
4259
Beilstein/REAXYS Number:
1731687
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.13

biological source

synthetic

assay

98%

refractive index

n20/D 1.4520 (lit.)

bp

173-176 °C/765 mmHg (lit.)

density

0.844 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

sulfurous

SMILES string

CCCCCCCS

InChI

1S/C7H16S/c1-2-3-4-5-6-7-8/h8H,2-7H2,1H3

InChI key

VPIAKHNXCOTPAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

105.8 °F - closed cup

flash_point_c

41 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alif Chebbi et al.
FEMS microbiology letters, 362(14) (2015-06-19)
Hydrogen sulfide (H2S) and thiols (RSH) generated by the phosphate industry cause harmful effects on human health and quality of life. The present study aims to investigate and evaluate a bacterial strain CAT37 isolated from gas-washing wastewaters in terms of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service