Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W421906

Sigma-Aldrich

Cornmint oil

Certified organic (NOP)

Synonym(s):

Mentha arvensis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
4219
MDL number:
UNSPSC Code:
12164502
organoleptic:
minty; camphoraceous
grade:
Certified organic (NOP)
Halal
Kosher
food allergen:
no known allergens

grade

Certified organic (NOP)
Halal
Kosher

origin

India origin

refractive index

n20/D 1.460 (lit.)

bp

218 °C/760 mmHg (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

minty; camphoraceous

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Legal Information

Certified organic by MOSA

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

150.0 °F - closed cup

flash_point_c

65.56 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service