Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W393600

Sigma-Aldrich

cis-3-Hexenyl valerate

mixture of isomers, natural (US), ≥97%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20O2
CAS Number:
Molecular Weight:
184.28
FEMA Number:
3936
EC Number:
Council of Europe no.:
10686
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.571
organoleptic:
green; tropical; fruity
grade:
FG
Fragrance grade
Kosher
natural (US)
biological source:
botanical
agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

botanical

grade

FG
Fragrance grade
Kosher
natural (US)

agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

assay

≥97%

refractive index

n20/D 1.435

density

0.882 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

organoleptic

green; tropical; fruity

SMILES string

CCCCC(=O)OCC\C=C/CC

InChI

1S/C11H20O2/c1-3-5-7-8-10-13-11(12)9-6-4-2/h5,7H,3-4,6,8-10H2,1-2H3/b7-5-

InChI key

XPFTVTFOOTVHIA-ALCCZGGFSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

170.0 °F

flash_point_c

76.67 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service