Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W363928

Sigma-Aldrich

β-Cyclocitral

≥95%

Synonym(s):

2,6,6-Trimethyl-1-cyclohexene-1-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
FEMA Number:
3639
EC Number:
Council of Europe no.:
10326
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.121
organoleptic:
tropical
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Halal
Kosher

assay

≥95%

refractive index

n20/D 1.497 (lit.)

bp

62-63 °C/3 mmHg (lit.)

density

0.943 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

tropical

SMILES string

[H]C(=O)C1=C(C)CCCC1(C)C

InChI

1S/C10H16O/c1-8-5-4-6-10(2,3)9(8)7-11/h7H,4-6H2,1-3H3

InChI key

MOQGCGNUWBPGTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Cyclocitral is a metabolite of the cyanobacteria of the genus Microcystis, which has a characteristic tobacco flavor.[1][2] It is one of the predominant volatile terpenoid compounds found in algal bloom water.[3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of Microcystis strains by alkyl sulfides and ?-cyclocitral.
Jiittner F
Zeitschrift fur Naturforschung C, 39(9-10), 867-871 (1984)
Analysis of volatile compounds in Algal bloom water by gas chromatography-mass spectrometry and sniffing
Yu RP, et al.
Fenxi Huaxue, 38, 1349-1352 (2010)
?-Cyclocitral and alkanes in Microcystis (Cyanophyceae).
Juttner F
Zeitschrift fur Naturforschung C, 31(9-10), 491-495 (1976)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service