Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W351105

Sigma-Aldrich

2-Methyl-4-pentenoic acid

≥98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2CH(CH3)CO2H
CAS Number:
Molecular Weight:
114.14
FEMA Number:
3511
EC Number:
Council of Europe no.:
10148
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.059
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

assay

≥98%

contains

0.10% alpha-tocopherol, synthetic as antioxidant

refractive index

n20/D 1.43 (lit.)

bp

195 °C (lit.)

density

0.949 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

cheese

SMILES string

CC(CC=C)C(O)=O

InChI

1S/C6H10O2/c1-3-4-5(2)6(7)8/h3,5H,1,4H2,2H3,(H,7,8)

InChI key

HVRZYSHVZOELOH-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service