Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W350303

Sigma-Aldrich

2-,3-,10-Mercaptopinane

mixture of isomers, FG

Synonym(s):

mango thiol, Pinanyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18S
CAS Number:
Molecular Weight:
170.31
FEMA Number:
3503
EC Number:
Council of Europe no.:
2332
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.035
NACRES:
NA.21

biological source

synthetic

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

refractive index

n20/D 1.5 (lit.)

bp

142 °C/20 mmHg (lit.)

density

0.98 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

green; tropical; sulfurous

SMILES string

CC1(C)[C@@H]2CCC(C)(S)[C@H]1C2

InChI

1S/C10H18S/c1-9(2)7-4-5-10(3,11)8(9)6-7/h7-8,11H,4-6H2,1-3H3/t7-,8+,10?/m1/s1

InChI key

ZPUCQOXKFCVYGL-HHCGNCNQSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

208.4 °F - closed cup

flash_point_c

98 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service