Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W349704

Sigma-Aldrich

cis-3-Hexenyl 2-methylbutanoate

≥97%, FCC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
184.28
FEMA Number:
3497
EC Number:
Council of Europe no.:
2345
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.854

biological source

synthetic

grade

Kosher

reg. compliance

FCC

assay

≥97%

refractive index

n20/D 1.432 (lit.)

density

0.878 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

apple; minty; waxy; herbaceous; pepper; sweet

SMILES string

[H]\C(CC)=C(/[H])CCOC(=O)C(C)CC

InChI

1S/C11H20O2/c1-4-6-7-8-9-13-11(12)10(3)5-2/h6-7,10H,4-5,8-9H2,1-3H3/b7-6-

InChI key

JKKGTSUICJWEKB-SREVYHEPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 1-10 ppm

Other Notes

Natural occurrence: Apricot, spearmint, black and green tea, fresh plums.

pictograms

Environment

hcodes

pcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

181.4 °F - closed cup

flash_point_c

83 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 25, 71-71 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service