Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W342718

Sigma-Aldrich

2,4-Dimethylbenzaldehyde

≥90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3CHO
CAS Number:
Molecular Weight:
134.18
FEMA Number:
3427
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.110
NACRES:
NA.21
organoleptic:
almond; cherry; spicy; sweet
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

assay

≥90%

refractive index

n20/D 1.549 (lit.)

bp

102-103 °C/14 mmHg (lit.)

mp

−9 °C (lit.)

density

0.962 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

almond; cherry; spicy; sweet

SMILES string

Cc1ccc(C=O)c(C)c1

InChI

1S/C9H10O/c1-7-3-4-9(6-10)8(2)5-7/h3-6H,1-2H3

InChI key

GISVICWQYMUPJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

192.2 °F - closed cup

flash_point_c

89 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias Onghena et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 31(12), 2090-2102 (2014-11-20)
In 2011, the European Union prohibited the production of polycarbonate (PC) baby bottles due to the toxic effects of the PC monomer bisphenol-A. Therefore, baby bottles made of alternative materials, e.g. polypropylene (PP) or polyethersulphone (PES), are currently marketed. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service