Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W335800

Sigma-Aldrich

2-Methoxy-3(5 or 6)-isopropylpyrazine

≥98%

Synonym(s):

3,5 Or 6-Isopropyl-2-methoxypyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2O
CAS Number:
Molecular Weight:
152.19
FEMA Number:
3358
EC Number:
Council of Europe no.:
11344
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.121

biological source

synthetic

grade

Halal
Kosher

assay

≥98%

refractive index

n20/D 1.495 (lit.)

density

0.996 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

green; horseradish; earthy; peanut; pepper

SMILES string

COc1cnc(cn1)C(C)C.COc2cncc(n2)C(C)C.COc3nccnc3C(C)C

InChI

1S/3C8H12N2O/c1-6(2)7-4-10-8(11-3)5-9-7;1-6(2)7-4-9-5-8(10-7)11-3;1-6(2)7-8(11-3)10-5-4-9-7/h3*4-6H,1-3H3

InChI key

MQHZLANELJKBBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 0.05 to 1.0 ppm

Other Notes

Natural occurrence: Green peas and potatoes.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

152.6 °F

flash_point_c

67 °C

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 103-103 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service