All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H12O2
CAS Number:
Molecular Weight:
152.19
FEMA Number:
3307
EC Number:
Council of Europe no.:
10355
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.058
biological source
synthetic
assay
≥99%
refractive index
n20/D 1.472 (lit.)
bp
93-94 °C/14 mmHg (lit.)
density
1.009 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
documentation
see Safety & Documentation for available documents
food allergen
no known allergens
organoleptic
vegetable; fruity
SMILES string
CC(CC=O)c1ccc(C)o1
InChI
1S/C9H12O2/c1-7(5-6-10)9-4-3-8(2)11-9/h3-4,6-7H,5H2,1-2H3
InChI key
DFSVNSCDOZSUCT-UHFFFAOYSA-N
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service