Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W330418

Sigma-Aldrich

3-Methyl-2-butanethiol

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(SH)CH3
CAS Number:
Molecular Weight:
104.21
FEMA Number:
3304
EC Number:
Council of Europe no.:
11510
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.049
NACRES:
NA.21
organoleptic:
sulfurous
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Halal
Kosher

assay

≥98%

refractive index

n20/D 1.444 (lit.)

bp

109-112 °C (lit.)

density

0.841 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

sulfurous

SMILES string

CC(C)C(C)S

InChI

1S/C5H12S/c1-4(2)5(3)6/h4-6H,1-3H3

InChI key

BFLXFRNPNMTTAA-UHFFFAOYSA-N

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

60.0 °F - closed cup

flash_point_c

15.56 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anamika Sharma et al.
Frontiers in chemistry, 6, 516-516 (2018-11-18)
The study involves exploring the three orthogonal sites for aromatic nucleophilic substitution in cyanuric chloride (TCT). The preferential order of incorporation of different nucleophiles (such as alcohol, thiol, and amine) was addressed both experimentally and theoretically. The preferential order for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service