Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W328400

Sigma-Aldrich

1-Furfurylpyrrole

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO
CAS Number:
Molecular Weight:
147.17
FEMA Number:
3284
EC Number:
Council of Europe no.:
2317
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Flavis number:
13.134

biological source

synthetic

grade

Halal
Kosher

vapor density

>1 (vs air)

assay

≥98%

refractive index

n20/D 1.531 (lit.)
n20/D 1.531

bp

76-78 °C/1 mmHg (lit.)

density

1.081 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

green; earthy; vegetable

SMILES string

C(c1ccco1)n2cccc2

InChI

1S/C9H9NO/c1-2-6-10(5-1)8-9-4-3-7-11-9/h1-7H,8H2

InChI key

BTBFUBUCCJKJOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

199.4 °F - closed cup

flash_point_c

93 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service