Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W326208

Sigma-Aldrich

Cyclopentanethiol

≥97%, FG

Synonym(s):

Cyclopentyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9SH
CAS Number:
Molecular Weight:
102.20
FEMA Number:
3262
EC Number:
Council of Europe no.:
2321
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.029
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

vapor density

0.96 (vs air)

vapor pressure

1 mmHg ( 20 °C)

assay

≥97%

refractive index

n20/D 1.4902 (lit.)

bp

129-131 °C/745 mmHg (lit.)

density

0.955 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

garlic; horseradish; onion; vegetable; alliaceous

SMILES string

SC1CCCC1

InChI

1S/C5H10S/c6-5-3-1-2-4-5/h5-6H,1-4H2

InChI key

WVDYBOADDMMFIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service