Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W318904

Sigma-Aldrich

2-Methyl-3(5 or 6)-furfurylthiopyrazine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2OS
CAS Number:
Molecular Weight:
206.26
FEMA Number:
3189
UNSPSC Code:
12164502
Flavis number:
13.151
NACRES:
NA.21
organoleptic:
coffee; nutty; roasted; sulfurous
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

assay

97%

refractive index

n20/D 1.600

density

1.210 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

coffee; nutty; roasted; sulfurous

InChI

1S/C10H10N2OS/c1-8-10(12-5-4-11-8)14-7-9-3-2-6-13-9/h2-6H,7H2,1H3

InChI key

PFRSWMCUERVSAT-UHFFFAOYSA-N

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

134.1 °F

flash_point_c

56.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service