Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W314218

Sigma-Aldrich

3,7-Dimethyl-6-octenoic acid

≥95%, FG

Synonym(s):

Citronellic acid, 3,7-Dimethyl-6-octenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CO2H
CAS Number:
Molecular Weight:
170.25
FEMA Number:
3142
EC Number:
Council of Europe no.:
616
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.036
organoleptic:
fatty; grassy; green; floral; smoky
grade:
FG
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012

assay

≥95%

refractive index

n20/D 1.454 (lit.)

bp

121-122 °C/1 mmHg (lit.)

density

0.923 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

fatty; grassy; green; floral; smoky

SMILES string

CC(CC\C=C(\C)C)CC(O)=O

InChI

1S/C10H18O2/c1-8(2)5-4-6-9(3)7-10(11)12/h5,9H,4,6-7H2,1-3H3,(H,11,12)

InChI key

GJWSUKYXUMVMGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Dermal - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service