Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W306540

Sigma-Aldrich

Thyme oil

white, Certified organic (NOP)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
3065
MDL number:
UNSPSC Code:
12164502

grade

Certified organic (NOP)
Halal
Kosher

origin

Albania origin

refractive index

n20/D 1.506

density

0.941 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

thyme

Looking for similar products? Visit Product Comparison Guide

Legal Information

Certified organic by MOSA

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

140.0 °F - closed cup

flash_point_c

60 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingming Guo et al.
Ultrasonics sonochemistry, 66, 104988-104988 (2020-03-31)
Essential oil nanoemulsions have been proven to have stronger antimicrobial effects compared to the essential oil alone or coarse emulsion. Sonoporation could be the promising candidate to trigger a synergistic effect with thyme essential oil nanoemulsion (TEON) and produce a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service