Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W306401

Sigma-Aldrich

Thyme oil

red

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
3064
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

synthetic (5% linalool added)

Quality Level

grade

Fragrance grade
Halal
Kosher

agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

optical activity

[α]20/D −1.8°, neat

origin

Spain origin

storage condition

protect from light

refractive index

n20/D 1.5

bp

190 °C

density

0.916 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

linalool, limonene (sum of D, L and DL), myrcene

organoleptic

herbaceous; camphoraceous; phenolic

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurrence: Thyme.

Biochem/physiol Actions

Taste at 50 ppm

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

140.0 °F - closed cup

flash_point_c

60 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mosciano, G.
Perfumer & Flavorist, 21, 54-54 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service