Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W306002

Sigma-Aldrich

Tetrahydrolinalool

≥97%, FCC

Synonym(s):

3,7-Dimethyl-3-octanol, Tetrahydrolinalool

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CH(CH2)3C(CH3)(OH)C2H5
CAS Number:
Molecular Weight:
158.28
FEMA Number:
3060
EC Number:
Council of Europe no.:
77
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.028
NACRES:
NA.21
organoleptic:
floral
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

reg. compliance

FCC
FDA 21 CFR 172.515

assay

≥97%

refractive index

n20/D 1.434 (lit.)

bp

71-73 °C/6 mmHg (lit.)

density

0.826 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

floral

SMILES string

CCC(C)(O)CCCC(C)C

InChI

1S/C10H22O/c1-5-10(4,11)8-6-7-9(2)3/h9,11H,5-8H2,1-4H3

InChI key

DLHQZZUEERVIGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

168.8 °F - closed cup

flash_point_c

76 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service