Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W303216

Sigma-Aldrich

Spearmint oil

terpeneless

Synonym(s):

Mentha spicata

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
3032
MDL number:
UNSPSC Code:
12164502

grade

Kosher

optical activity

[α]20/D −58°, neat

composition

Carvone, 55-80% (typical)

origin

USA origin

refractive index

n20/D 1.489 (lit.)

bp

228 °C (lit.)

density

0.941 g/mL at 25 °C

application(s)

flavors and fragrances

organoleptic

green; camphoraceous; minty

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 10 ppm

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

201.2 °F - closed cup

flash_point_c

94 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 27, 92-92 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service