Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W299219

Sigma-Aldrich

Rosemary oil, Spanish

Sign Into View Organizational & Contract Pricing


About This Item

FEMA Number:
2992
UNSPSC Code:
12164502
organoleptic:
herbaceous
grade:
Kosher
biological source:
Rosemarinus officinalis L.
Pricing and availability is not currently available.

biological source

Rosemarinus officinalis L.

Quality Level

grade

Kosher

origin

Spain origin

refractive index

n20/D 1.468

bp

148.5 °C

density

0.903 g/mL at 25 °C

application(s)

flavors and fragrances

organoleptic

herbaceous

Preparation Note

Extraction method: steam distillation

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

120.2 °F - closed cup

flash_point_c

49 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service