Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W286508

Sigma-Aldrich

Phenethyl 2-furoate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12O3
CAS Number:
Molecular Weight:
216.23
FEMA Number:
2865
Council of Europe no.:
362
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.006

biological source

synthetic

grade

Kosher

assay

≥98%

refractive index

n20/D 1.546 (lit.)

density

1.139 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

organoleptic

musty

SMILES string

O=C(OCCc1ccccc1)c2ccco2

InChI

1S/C13H12O3/c14-13(12-7-4-9-15-12)16-10-8-11-5-2-1-3-6-11/h1-7,9H,8,10H2

InChI key

QKPSYARWSBJEDY-UHFFFAOYSA-N

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service