Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W286303

Sigma-Aldrich

Phenethyl cinnamate

≥96%, FG

Synonym(s):

Benzyl carbinyl cinnamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCO2CH2CH2C6H5
CAS Number:
Molecular Weight:
252.31
FEMA Number:
2863
EC Number:
Council of Europe no.:
336
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.743
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

assay

≥96%

mp

54-56 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

balsamic

SMILES string

O=C(OCCc1ccccc1)\C=C\c2ccccc2

InChI

1S/C17H16O2/c18-17(12-11-15-7-3-1-4-8-15)19-14-13-16-9-5-2-6-10-16/h1-12H,13-14H2/b12-11+

Inchi Key

MJQVZIANGRDJBT-VAWYXSNFSA-N

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luana Chiquetto Paracatu et al.
Evidence-based complementary and alternative medicine : eCAM, 2014, 793629-793629 (2014-09-26)
Numerous anti-inflammatory properties have been attributed to caffeic acid phenethyl ester (CAPE), an active component of propolis. NADPH oxidases are multienzymatic complexes involved in many inflammatory diseases. Here, we studied the importance of the CAPE hydrophobicity on cell-free antioxidant capacity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service