W284505
Black pepper oil
natural
Synonym(s):
Piper Nigrum
About This Item
Recommended Products
biological source
Piper nigrum L.
grade
Fragrance grade
Kosher
natural
agency
follows IFRA guidelines
reg. compliance
EU Regulation 1223/2009
optical activity
[α]20/D −25°, neat
origin
India origin
refractive index
n20/D 1.483 (lit.)
bp
166 °C (lit.)
density
0.873 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
organoleptic
spicy; pepper; warm
Biochem/physiol Actions
Preparation Note
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
129.2 °F - closed cup
flash_point_c
54 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service