Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W277509

Sigma-Aldrich

Neryl isobutyrate

≥97%, stabilized, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
224.34
FEMA Number:
2775
EC Number:
Council of Europe no.:
299
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.424
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

assay

≥97%

contains

α-tocopherol as stabilizer

refractive index

n20/D 1.457 (lit.)

bp

229 °C (lit.)

density

0.895 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

fresh; fruity; raspberry; strawberry; sweet

SMILES string

CC(C)C(=O)OC\C=C(\C)CC\C=C(/C)C

InChI

1S/C14H24O2/c1-11(2)7-6-8-13(5)9-10-16-14(15)12(3)4/h7,9,12H,6,8,10H2,1-5H3/b13-9-

Inchi Key

OGJYXQFXLSCKTP-LCYFTJDESA-N

General description

Neryl isobutyrate has been reported in the essential oils of plants such as Artemisia absinthium L.[1] and Telekia speciosa (Schreb.) Baumg.[2]

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatiles of Telekia speciosa (Schreb.) Baumg.(Asteraceae) from Serbia.
Radulovic N, et al.
J. Essent. Oil Res., 22(3), 250-254 (2010)
Composition of the essential oil of Artemisia absinthium L. of different geographical origin. Proceedings-estonian Academy of Sciences
Orav A, et al.
Proceedings of the Estonian Academy of Sciences, 55(3), 155-155 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service