Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W277401

Sigma-Aldrich

Neryl butyrate

≥90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
224.34
FEMA Number:
2774
EC Number:
Council of Europe no.:
505
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.167
NACRES:
NA.21

biological source

synthetic

Quality Level

assay

≥90%

refractive index

n20/D 1.459 (lit.)

bp

240 °C (lit.)

density

0.898 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

fatty; green; leafy; oily; orange; waxy; sweet

SMILES string

CCCC(=O)OC\C=C(\C)CC\C=C(\C)C

InChI

1S/C14H24O2/c1-5-7-14(15)16-11-10-13(4)9-6-8-12(2)3/h8,10H,5-7,9,11H2,1-4H3/b13-10-

InChI key

ZSBOMYJPSRFZAL-RAXLEYEMSA-N

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service