Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W265734

Sigma-Aldrich

Mandarin green oil

Certified organic (NOP)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2657
MDL number:
UNSPSC Code:
12164502
organoleptic:
orange
grade:
Certified organic (NOP)
Halal
Kosher
biological source:
Citrus reticulata
food allergen:
no known allergens

biological source

Citrus reticulata

grade

Certified organic (NOP)
Halal
Kosher

reg. compliance

FDA 21 CFR 117

origin

Italy origin

refractive index

n20/D 1.474

density

0.852 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

orange

Legal Information

Certified organic by MOSA

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

133.0 °F - closed cup

flash_point_c

56.11 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service