Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W265101

Sigma-Aldrich

Lovage oil

natural

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2651
MDL number:
UNSPSC Code:
12164502

biological source

Levisticum officinale Koch

grade

Kosher
natural

optical activity

[α]20/D +2°, neat

origin

France origin

refractive index

n20/D 1.546 (lit.)

density

1.036 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

cheese; green; meaty; fruity; earthy; vegetable; sweet; walnut

Biochem/physiol Actions

Odor at 1.0%
Taste at 10 ppm

Other Notes

Natural occurrence: Lovage root.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 27, 92-92 (2002)
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 77-77 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service